4-Chloro-3-(dimethylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Chloro-3-(dimethylcarbamoyl)phenyl)boronic acid

A137137-1G A137137-250MG A137137-5G
76782-712EA 146.41 USD
76782-712 76782-714 76782-716
4-Chloro-3-(dimethylcarbamoyl)phenylboronic acid 98%
4-Chloro-3-(dimethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BClNO₃
MW: 227.4500 g/mol
MDL Number: MFCD07783868
CAS Number: 871332-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR