(S)-2-(4-(tert-Butoxycarbonyl)morpholin-2-yl)acetic acid 95%

Supplier: Ambeed
Warning

A136211-250MG A136211-5G A136211-100MG A136211-1G
76782-666EA 46 USD
76782-666 77566-704 77599-976 77599-978
(S)-2-(4-(tert-Butoxycarbonyl)morpholin-2-yl)acetic acid 95%
(S)-2-(4-(tert-Butoxycarbonyl)morpholin-2-yl)acetic acid

Precaución: For research use only.

Formula: C₁₁H₁₉NO₅
MDL Number: MFCD13189487
CAS Number: 1257850-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR