4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: trans,trans-4'-Pentylbicyclohexyl-4-carboxylic acid 4-Fluorophenyl ester, 4-Fluorophenyl trans,trans-4'-Pentylbicyclohexyl-4-carboxylate, (Trans,trans)-4-fluorophenyl 4'-pentyl-[1,1'-bi(cyclohexane)]-4-carboxylate

A135885-1G A135885-5G
76782-654EA 11.67 USD
76782-654 76782-656
4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate 97%
4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate

Precaución: for Research Use Only

Formula: C₂₄H₃₅FO₂
MW: 374.54 g/mol
MDL Number: MFCD11053423
CAS Number: 88878-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR