Tris(2-butoxyethyl)phosphate 98%

Supplier: Ambeed
Warning

Synonyms: Tris(2-butoxyethyl) phosphate, Phosphoric Acid Tris(2-butoxyethyl) Ester

A135835-1000G A135835-100G A135835-500G
76782-648EA 118.83 USD
76782-648 76782-650 76782-652
Tris(2-butoxyethyl)phosphate 98%
Tris(2-butoxyethyl)phosphate

Precaución: for Research Use Only

Formula: C₁₈H₃₉O₇P
MW: 398.48 g/mol
MDL Number: MFCD00009456
CAS Number: 78-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR