Triethylene glycol diacetate 98%

Supplier: Ambeed
Warning

Synonyms: Triethylene Glycol Diacetate

A135212-100G A135212-25G
76782-618EA 32 USD
76782-618 76782-620
Triethylene glycol diacetate 98%
Triethylene glycol diacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₆
MW: 234.25 g/mol
MDL Number: MFCD00026213
CAS Number: 111-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR