4-Iodoisoquinoline 97%

Supplier: Ambeed
Warning

A135119-100MG A135119-250MG A135119-1G A135119-5G
76782-614EA 18 USD
76782-614 76782-616 77666-952 77666-954
4-Iodoisoquinoline 97%
4-Iodoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₆IN
MW: 255.0600 g/mol
MDL Number: MFCD00234496
CAS Number: 55270-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR