tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Boc-DL-Prolinol, tert-Butyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate

A135106-100G A135106-10G A135106-1G A135106-25G A135106-5G
76782-604EA 247 USD
76782-604 76782-606 76782-608 76782-610 76782-612
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate 97%
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD01456556
CAS Number: 170491-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR