Methyl-6-chloropyrazine-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 6-chloropyrazine-2-carboxylate
A134965-100G
A134965-1G
A134965-250MG
A134965-25G
A134965-5G
76782-594EA
471.04
USD
76782-594
76782-596
76782-598
76782-600
76782-602
Methyl-6-chloropyrazine-2-carboxylate 97%
Methyl-6-chloropyrazine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₅ClN₂O₂ MW: 172.57 g/mol |
MDL Number:
MFCD10686598 CAS Number: 23611-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...