N-Acetyl-D-lactosamine 98%

Supplier: Ambeed
Warning

Synonyms: N-Acetyllactosamine, 2-(Acetylamino)-2-deoxy-4-O-β-D-galactopyranosyl-D-glucose, N-((2R,3R,4S,5R)-3,5,6-Trihydroxy-1-oxo-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexan-2-yl)acetamide

A134794-100MG A134794-50MG A134794-10MG
76782-584EA 526.21 USD
76782-584 77599-948 77666-948
N-Acetyl-D-lactosamine 98%
N-Acetyl-D-lactosamine

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₁₁
MW: 383.35 g/mol
MDL Number: MFCD00063785
CAS Number: 32181-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR