N-Boc-4-bromo-2-methylaniline 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-bromo-2-methylphenyl)carbamate, tert-Butyl 4-bromo-2-methylphenylcarbamate, 4-Bromo-2-methylaniline, N-BOC protected

A134429-1G A134429-250MG A134429-5G
76782-570EA 20.16 USD
76782-570 76782-572 76782-574
N-Boc-4-bromo-2-methylaniline 97%
N-Boc-4-bromo-2-methylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂
MW: 286.1700 g/mol
MDL Number: MFCD01765429
CAS Number: 306937-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR