trans-4-Aminocyclohexanecarboxylic acid hydrochloride 96%

Supplier: Ambeed
Warning

A134349-100G A134349-10G A134349-1G A134349-25G A134349-5G
76782-562EA 708.69 USD
76782-562 76782-564 76782-566 76782-568 77666-944
trans-4-Aminocyclohexanecarboxylic acid hydrochloride 96%
trans-4-Aminocyclohexanecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
MW: 179.65 g/mol
MDL Number: MFCD03844587
CAS Number: 27960-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR