4-Amino-6-bromopyrimidine 98%

Supplier: Ambeed
Warning

A134341-100MG A134341-10G A134341-1G A134341-250MG A134341-5G A134341-25G
76782-552EA 5 USD
76782-552 76782-554 76782-556 76782-558 76782-560 77871-638
4-Amino-6-bromopyrimidine 98%
4-Amino-6-bromopyrimidine

Precaución: for Research Use Only

Formula: C₄H₄BrN₃
MDL Number: MFCD00233963
CAS Number: 1159818-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR