2-Chloro-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride 98%

Supplier: Ambeed
Warning

A134174-100MG A134174-10G A134174-1G A134174-250MG A134174-25G A134174-5G
76782-534EA 6 USD
76782-534 76782-536 76782-538 76782-540 76782-542 76782-544
2-Chloro-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride 98%
2-Chloro-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀Cl₂N₂
MDL Number: MFCD09835503
CAS Number: 766545-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR