Phenanthrene-9,10-diamine 95%

Supplier: Ambeed
Warning

A134010-100MG A134010-1G A134010-250MG
76782-518EA 35 USD
76782-518 76782-520 77599-928
Phenanthrene-9,10-diamine 95%
Phenanthrene-9,10-diamine

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂
MDL Number: MFCD00001178
CAS Number: 53348-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR