Boc-N-ME-PHG-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-N-methyl-L-phenylglycine, Boc-N-Me-Phg-OH

A133537-1G A133537-5G
76782-492EA 16 USD
76782-492 76782-494
Boc-N-ME-PHG-OH 98%
Boc-N-ME-PHG-OH

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.309 g/mol
MDL Number: MFCD00038799
CAS Number: 30925-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR