(R)-1-((Benzyloxy)carbonyl)piperidine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A133393-1G A133393-250MG A133393-25G
76782-484EA 28.65 USD
76782-484 76782-486 76782-488
(R)-1-((Benzyloxy)carbonyl)piperidine-3-carboxylic acid 98%
(R)-1-((Benzyloxy)carbonyl)piperidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MW: 263.2900 g/mol
MDL Number: MFCD04114972
CAS Number: 160706-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR