4-Methyl-1H-benzimidazole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A133313-1G A133313-250MG A133313-5G
76782-478EA 72 USD
76782-478 76782-480 76782-482
4-Methyl-1H-benzimidazole-2-carboxylic acid 95%
4-Methyl-1H-benzimidazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD09751735
CAS Number: 673487-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR