Ethyl-4-formyl-1H-pyrrole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-formylpyrrole-2-carboxylate, Ethyl 4-formyl-1H-pyrrole-2-carboxylate

A132968-10G A132968-250MG A132968-25G A132968-5G
76782-458EA 141 USD
76782-458 76782-460 76782-462 76782-464
Ethyl-4-formyl-1H-pyrrole-2-carboxylate 97%
Ethyl-4-formyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD04122505
CAS Number: 7126-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR