Z-Phe-ala 95%

Supplier: Ambeed
Warning

Synonyms: Z-Phe-Ala-OH

A132900-10G A132900-1G A132900-250MG A132900-25G A132900-5G
76782-448EA 106.09 USD
76782-448 76782-450 76782-452 76782-454 76782-456
Z-Phe-ala 95%
Z-Phe-ala

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₂O₅
MW: 370.405 g/mol
MDL Number: MFCD00037237
CAS Number: 21881-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR