1-(4-Chloro-3-nitrobenzenesulfonyl)-1,2,3,4-tetrahydroquinoline 98%

Supplier: Ambeed
Warning

A132873-10G A132873-250MG A132873-25G
76782-442EA 132.62 USD
76782-442 76782-444 76782-446
1-(4-Chloro-3-nitrobenzenesulfonyl)-1,2,3,4-tetrahydroquinoline 98%
1-(4-Chloro-3-nitrobenzenesulfonyl)-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClN₂O₄S
MW: 352.7900 g/mol
MDL Number: MFCD08275686
CAS Number: 847170-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR