6-Bromopyridine-2-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A132245-100MG A132245-1G A132245-250MG A132245-5G
76782-410EA 46 USD
76782-410 76782-412 76782-414 77871-620
6-Bromopyridine-2-sulfonyl chloride 95%
6-Bromopyridine-2-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₅H₃BrClNO₂S
MDL Number: MFCD11870765
CAS Number: 912934-77-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR