5-Fluoro-2-methoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (5-Fluoro-2-methoxyphenyl)methanol

A132217-1G A132217-25G A132217-5G
76782-408EA 17 USD
76782-408 77871-616 77871-617
5-Fluoro-2-methoxybenzyl alcohol 98%
5-Fluoro-2-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MDL Number: MFCD00671767
CAS Number: 426831-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR