5-Bromo-2,4-difluorotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,4-difluoro-5-methylbenzene

A131539-1G A131539-25G A131539-5G
76782-386EA 6.37 USD
76782-386 76782-388 76782-390
5-Bromo-2,4-difluorotoluene 98%
5-Bromo-2,4-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
MDL Number: MFCD15144620
CAS Number: 159277-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR