(trans,trans)-4-Pentyl-4'-vinyl-1,1'-bi(cyclohexane) 98%

Supplier: Ambeed
Warning

A131416-1G A131416-25G A131416-5G
76782-380EA 14 USD
76782-380 76782-382 76782-384
(trans,trans)-4-Pentyl-4'-vinyl-1,1'-bi(cyclohexane) 98%
(trans,trans)-4-Pentyl-4'-vinyl-1,1'-bi(cyclohexane)

Precaución: for Research Use Only

Formula: C₁₉H₃₄
MW: 262.47 g/mol
MDL Number: MFCD11053467
CAS Number: 129738-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR