4-bromo-1-methyl-1H-pyrazole-3-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile

A131264-1G A131264-250MG A131264-5G
76782-368EA 98.67 USD
76782-368 76782-370 76782-372
4-bromo-1-methyl-1H-pyrazole-3-carbonitrile 98%
4-bromo-1-methyl-1H-pyrazole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄BrN₃
MW: 186.012 g/mol
MDL Number: MFCD00103545
CAS Number: 287922-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR