8-((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one 98%

Supplier: Ambeed
Warning

A131116-10MG A131116-1MG A131116-5MG
76782-362EA 47.75 USD
76782-362 76782-364 76782-366
8-((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one 98%
8-((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₇H₃₀O₁₄
MW: 578.52 g/mol
MDL Number: MFCD29913069
CAS Number: 64820-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR