Tyrphostin B46 98%

Supplier: Ambeed
Warning

Synonyms: (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)prop-2-enamide, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)-2-propenamide, AG 555, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)acrylamide, Tyrphostin AG-555

A131103-100MG A131103-10MG A131103-1MG A131103-50MG A131103-5MG
76782-352EA 179.3 USD
76782-352 76782-354 76782-356 76782-358 76782-360
Tyrphostin B46 98%
Tyrphostin B46

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₂O₃
MW: 322.36 g/mol
MDL Number: MFCD00209865
CAS Number: 133550-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR