trans-4-(trans-4-Propylcyclohexyl)cyclohexanol 97%

Supplier: Ambeed
Warning

A131072-100G A131072-10G A131072-1G A131072-25G A131072-5G
76782-340EA 102 USD
76782-340 76782-342 76782-344 76782-346 76782-348
trans-4-(trans-4-Propylcyclohexyl)cyclohexanol 97%
trans-4-(trans-4-Propylcyclohexyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₅H₂₈O
MDL Number: MFCD09752068
CAS Number: 82832-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR