4-Bromo-2-chloro-3-fluorophenol 95%

Supplier: Ambeed
Warning

A130998-100MG A130998-5G A130998-250MG
76782-336EA 27.59 USD
76782-336 76782-338 77666-876
4-Bromo-2-chloro-3-fluorophenol 95%
4-Bromo-2-chloro-3-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrClFO
MW: 225.44 g/mol
MDL Number: MFCD11849932
CAS Number: 1233026-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR