4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine 97%

Supplier: Ambeed
Warning

A130659-10G A130659-1G A130659-250MG
76782-318EA 901.77 USD
76782-318 76782-320 76782-322
4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine 97%
4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.6000 g/mol
MDL Number: MFCD00222101
CAS Number: 50432-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR