7-Aminoquinoline 97%

Supplier: Ambeed
Warning

Synonyms: Quinolin-7-amine

A129894-10G A129894-1G A129894-250MG A129894-25G A129894-5G
76782-284EA 157.02 USD
76782-284 76782-286 76782-288 76782-290 76782-292
7-Aminoquinoline 97%
7-Aminoquinoline

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18 g/mol
MDL Number: MFCD01646243
CAS Number: 580-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR