Succinylsulfathiazole 98%

Supplier: Ambeed
Warning

Synonyms: N4-Succinyl-N-(2-thiazolyl)sulphanilamide, 4-(Succinylamino)-N-(2-thiazolyl)benzenesulphonamide, 4-Oxo-4-((4-(N-(thiazol-2-yl)sulphamoyl)phenyl)amino)butanoic acid

A129827-100MG A129827-25G A129827-5G A129827-100G A129827-500G
76782-278EA 5.31 USD
76782-278 76782-280 76782-282 77666-854 77666-856
Succinylsulfathiazole 98%
Succinylsulphathiazole

Precaución: for Research Use Only

Formula: C₁₃H₁₃N₃O₅S₂
MW: 355.38 g/mol
MDL Number: MFCD00022437
CAS Number: 116-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR