2-Amino-4,5-difluorophenol 98%

Supplier: Ambeed
Warning

A129617-1G A129617-250MG
76782-262EA 141 USD
76782-262 77871-599
2-Amino-4,5-difluorophenol 98%
2-Amino-4,5-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₅F₂NO
MDL Number: MFCD03094185
CAS Number: 163734-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR