6-Bromo-2-chloroquinazoline 97%

Supplier: Ambeed
Warning

A129515-1G A129515-250MG A129515-25G A129515-5G
76782-244EA 19.1 USD
76782-244 76782-246 76782-248 76782-250
6-Bromo-2-chloroquinazoline 97%
6-Bromo-2-chloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂
MW: 243.4900 g/mol
MDL Number: MFCD09261000
CAS Number: 882672-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR