3-Bromo-9-phenyl-9H-carbazole 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-9-phenylcarbazole

A129393-100G A129393-10G A129393-1G A129393-25G A129393-5G
76782-230EA 142.17 USD
76782-230 76782-232 76782-234 76782-236 76782-238
3-Bromo-9-phenyl-9H-carbazole 97%
3-Bromo-9-phenyl-9H-carbazole

Precaución: for Research Use Only

Formula: C18H12BrN
MW: 322.20 g/mol
MDL Number: MFCD11977305
CAS Number: 1153-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR