7-Bromophthalazin-1(2H)-one 95%

Supplier: Ambeed
Warning

A129198-100MG A129198-1G A129198-250MG A129198-5G
76782-214EA 21 USD
76782-214 76782-216 76782-218 76782-220
7-Bromophthalazin-1(2H)-one 95%
7-Bromophthalazin-1(2H)-one

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
MDL Number: MFCD11518962
CAS Number: 152265-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR