tert-Butyl-5-nitro-1H-indole-1-carboxylate 95%

Supplier: Ambeed
Warning

A128882-1G A128882-250MG
76782-210EA 72 USD
76782-210 76782-212
tert-Butyl-5-nitro-1H-indole-1-carboxylate 95%
tert-Butyl-5-nitro-1H-indole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₄
MDL Number: MFCD09953094
CAS Number: 166104-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR