6-Methyl-2-(methylthio)-1H-pyrimidin-4-one 98%

Supplier: Ambeed
Warning

A128841-1G A128841-25G A128841-10G A128841-5G
76782-206EA 7 USD
76782-206 76782-208 77871-596 77871-597
6-Methyl-2-(methylthio)-1H-pyrimidin-4-one 98%
6-Methyl-2-(methylthio)-1H-pyrimidin-4-one

Precaución: for Research Use Only

Formula: C₆H₈N₂OS
MDL Number: MFCD01022058
CAS Number: 6328-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR