4-Bromo-1-(2-tetrahydropyranyl)-1H-pyrazole 97%

Supplier: Ambeed
Warning

A128833-10G A128833-1G A128833-250MG A128833-5G
76782-198EA 27 USD
76782-198 76782-200 76782-202 76782-204
4-Bromo-1-(2-tetrahydropyranyl)-1H-pyrazole 97%
4-Bromo-1-(2-tetrahydropyranyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O
MDL Number: MFCD12026052
CAS Number: 82099-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR