tert-Butyl-1H-pyrazole-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrazole, N1-BOC protected, tert-Butyl 1H-pyrazole-1-carboxylate

A128668-100G A128668-10G A128668-1G A128668-25G A128668-5G
76782-168EA 161.26 USD
76782-168 76782-170 76782-172 76782-174 76782-176
tert-Butyl-1H-pyrazole-1-carboxylate 95%
tert-Butyl-1H-pyrazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.1900 g/mol
MDL Number: MFCD00800729
CAS Number: 219580-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR