Z-D-Glu(OtBu)-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-D-glutamic-acid 5-tert-butyl ester

A128616-100G A128616-1G A128616-5G A128616-25G A128616-500G
76782-162EA 183.54 USD
76782-162 76782-164 76782-166 77599-876 77666-844
Z-D-Glu(OtBu)-OH 98%
Z-D-Glu(OtBu)-OH

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₆
MDL Number: MFCD00077018
CAS Number: 51644-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR