5-Bromo-N-methylnicotinamide 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-methyl-3-pyridinecarboxamide

A128379-1G A128379-250MG A128379-5G
76782-152EA 31.83 USD
76782-152 76782-154 76782-156
5-Bromo-N-methylnicotinamide 97%
5-Bromo-N-methylnicotinamide

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O
MW: 215.05 g/mol
MDL Number: MFCD01788369
CAS Number: 153435-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR