2-Chloro-5-iodo-4-methylpyridine 97%

Supplier: Ambeed
Warning

A127683-10G A127683-1G A127683-250MG A127683-25G A127683-5G
76782-112EA 209 USD
76782-112 76782-114 76782-116 76782-118 76782-120
2-Chloro-5-iodo-4-methylpyridine 97%
2-Chloro-5-iodo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅ClIN
MDL Number: MFCD13185454
CAS Number: 550347-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR