1-(5-Bromopyrimidin-2-yl)-4-piperidinol 97%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Bromopyrimidin-2-yl)piperidin-4-ol

A127206-1G A127206-250MG A127206-5G
76782-096EA 37.14 USD
76782-096 76782-098 76782-100
1-(5-Bromopyrimidin-2-yl)-4-piperidinol 97%
1-(5-Bromopyrimidin-2-yl)-4-piperidinol

Precaución: for Research Use Only

Formula: C₉H₁₂BrN₃O
MW: 258.12 g/mol
MDL Number: MFCD03646019
CAS Number: 887425-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR