6-Chloro-5-nitroquinoline 98%

Supplier: Ambeed
Warning

A126769-100MG A126769-10G A126769-1G A126769-250MG
76782-074EA 5.31 USD
76782-074 76782-076 76782-078 76782-080
6-Chloro-5-nitroquinoline 98%
6-Chloro-5-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClN₂O₂
MW: 208.6012 g/mol
MDL Number: MFCD00093616
CAS Number: 86984-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR