Vandetanib 98%

Supplier: Ambeed
Warning

Synonyms: ZD6474, N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-((1-methylpiperidin-4-yl)methoxy)quinazolin-4-amine

A126372-1MG A126372-25MG A126372-5MG A126372-250MG A126372-25G
76782-038EA 5 USD
76782-038 76782-040 76782-042 77871-573 77871-574
Vandetanib 98%
Vandetanib

Precaución: for Research Use Only

Formula: C₂₂H₂₄BrFN₄O₂
MW: 475.4 g/mol
MDL Number: MFCD07772346
CAS Number: 443913-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR