3-Iodo-6-methoxy-1H-indazole 97%

Supplier: Ambeed
Warning

A126270-100MG A126270-1G A126270-250MG
76782-026EA 28 USD
76782-026 76782-028 76782-030
3-Iodo-6-methoxy-1H-indazole 97%
3-Iodo-6-methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇IN₂O
MDL Number: MFCD09266218
CAS Number: 936138-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR