2-Biphenylacetonitrile 96%

Supplier: Ambeed
Warning

Synonyms: 2-([1,1'-Biphenyl]-2-yl)acetonitrile

A126261-10G A126261-1G A126261-250MG
76782-020EA 255.68 USD
76782-020 76782-022 76782-024
2-Biphenylacetonitrile 96%
2-Biphenylacetonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
MW: 193.2400 g/mol
MDL Number: MFCD00060294
CAS Number: 19853-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR