N-Boc-m-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3-Aminophenyl)carbamate, (3-Aminophenyl)carbamic acid tert-Butyl ester, N-Boc-1,3-phenylenediamine, 3-(tert-Butoxycarbonylamino)aniline, N-(tert-Butoxycarbonyl)-1,3-phenylenediamine, tert-Butyl (3-aminophenyl)carbamate, 3-(Boc-amino)aniline

A125555-100G A125555-10G A125555-1G A125555-25G A125555-5G
76781-970EA 78 USD
76781-970 76781-972 76781-974 76781-976 76781-978
N-Boc-m-phenylenediamine 98%
N-Boc-m-phenylenediamine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD06657915
CAS Number: 68621-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR