6-Bromo-2-naphthyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

Synonyms: 6-Bromo-2-naphthyl Triflate, Trifluoromethanesulphonic acid 6-Bromo-2-naphthyl Ester, 6-Bromonaphthalen-2-yl trifluoromethanesulphonate

A125201-1G A125201-250MG A125201-25G
76781-954EA 28 USD
76781-954 76781-956 76781-958
6-Bromo-2-naphthyl trifluoromethanesulfonate 95%
6-Bromo-2-naphthyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₁H₆BrF₃O₃S
MW: 355.13 g/mol
MDL Number: MFCD03093642
CAS Number: 151600-02-1
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR